Learn More
Sino Biological Anti-FGF1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10013MM15100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human aFGF / FGF1 (rh aFGF / FGF1; Catalog#10013-HNAE; AAA79245.1; Phe16-Asp155). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1180