Learn More
Sino Biological Anti-FGF-10 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10573MM0420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FGF10 / KGF2 (rh FGF10 / KGF2; Catalog#10573-HNAE; NP_004456.1; Gln38-Ser208). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9680