Learn More
Sino Biological Anti-FGF-10 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10573T32100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FGF10 / KGF2 (rh FGF10 / KGF2; Catalog#10573-HNAE; NP_004456.1; Gln38-Ser208). FGF10 / KGF2 specific IgG was purified by Human FGF10 / KGF2 affinity chromatography.
Catalog No. 50-241-0920