Learn More
Sino Biological Recombinant Human FGF-18 Protein (His Tag)
Supplier:  Sino Biological 13206H08H1
Encompass_Preferred
A DNA sequence encoding the human FGF18 (O76093) (Met 1-Ala 207) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4277