Learn More
Bon Opus Biosciences Recombinant Human Fibroblast Growth Factor 19/FGF-19 (N-6His)
SDP
Supplier:  Bon Opus Biosciences CG7450UG
Encompass_Preferred
Recombinant Human Fibroblast growth factor 19 is produced by our E.coli expression system and the target gene encoding Phe27-Lys216 is expressed with a 6His tag at the N-terminus. Bon Opus Cat. #CG74
Catalog No. 50-726-562