Learn More
ACROBiosystems ACROBiosystems Human FGF19 Protein, Fc Tag
Supplier:  ACROBiosystems FG9H5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Human FGF19, Fc Tag (FG9-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Lys 216 (Accession # O95750-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 47.9 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6324