Learn More
ACROBiosystems ACROBiosystems Human FGF19 Protein, His Tag
Supplier:  ACROBiosystems FG9H51H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Human FGF19, His Tag (FG9-H51H3) is expressed from E.coli cells. It contains AA Leu 25 - Lys 216 (Accession # O95750-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.3 kDa.
Catalog No. 50-210-6322