Learn More
Abcam Human FGF2 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502656880
Encompass_Preferred
Human FGF2 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human FGF2.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6880 Supplier Abcam Supplier No. AB25667610X96TESTS
Human FGF2 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human FGF2.
The product is subject to the following: Abcam Restricted Use Statement