Learn More
Sino Biological Anti-basic FGF/FGF2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10014T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human bFGF/FGF2 (rh bFGF/FGF2; Catalog#10014-HNAE; NP_001997.5; Pro143-Ser288). bFGF/FGF2 specific IgG was purified by Human bFGF/FGF2 affinity chromatography.
Catalog No. 50-241-0588