Learn More
Sino Biological Recombinant Human KGF/FGF-7 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10210H07E20
Encompass_Preferred
A DNA sequence encoding the human KGF (P21781) (Cys32-Thr194) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-3171