Learn More
Sino Biological Recombinant Human KSP37/FGFBP2 Protein (His Tag)
Supplier:  Sino Biological 12139H08B100
Encompass_Preferred
A DNA sequence encoding the human FGFBP2 (NP_114156.1) (Met1-Gly223) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4455