Learn More
Sino Biological Human FGFBP3 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10986RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FGFBP3 (rh FGFBP3; Catalog#10986-H08B; Q8TAT2; Met1-Gly258). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-2423