Learn More
Sino Biological Anti-FGFR1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10616MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FGFR1 / CD331 (rh FGFR1 / CD331; Catalog#10616-H08H; NP_075594.1; Met1-Glu285). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8613