Learn More
Sino Biological Recombinant Human FGFR1 Protein (beta (IIIb), His Tag)
Supplier:  Sino Biological 16480H08H200
Encompass_Preferred
A DNA sequence encoding the human FGFR1 beta (IIIb) (NP_075594.1) (Met1-Lys221 and Ala270-Glu285 linked by the FGFR1 IIIb Ig domain (AAB19502.1) (His1-Pro47)) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4030