Learn More
Sino Biological Recombinant Human FGFR1 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10616H08HB20
Encompass_Preferred
A DNA sequence encoding the human FGFR1 (NP_075594.1) extracellular domain (Met1-Glu285) was expressed with C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5246