Learn More
Sino Biological Recombinant Human FGFR1 Protein (alpha (IIIb), His Tag), HPLC-verified
Supplier:  Sino Biological 16482H08H50
Encompass_Preferred
A DNA sequence encoding the human FGFR1 alpha (IIIb) (NP_056934.2) (Met1-Lys310 and Ala359-Glu374 linked by FGFR1 IIIb Ig domain (AAB19502.1) (His1-Pro47)) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4028