Learn More
Sino Biological Anti-FGFR2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10824MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FGFR2 / CD332 (rh FGFR2 / CD332; Catalog#10824-H08H; NP_000132.3; Met1-Glu377). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.
Catalog No. 50-255-5145