Learn More
Sino Biological Anti-FGFR2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10824T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FGFR2 / CD332 (rh FGFR2 / CD332; Catalog#10824-H20B1; NP_000132.3; Met400-Thr821). FGFR2 / CD332 specific IgG was purified by Human FGFR2 / CD332 affinity chromatography.
Catalog No. 50-255-5141