Learn More
Sino Biological Recombinant Human FGFR2 Protein (ECD, alpha(IIIb), His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 16485H49HB20
Encompass_Preferred
A DNA sequence encoding the Human FGFR2 (NP_075259.4)(Met1-Glu378) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-8188