Learn More
Sino Biological Recombinant Human FGFR3 Protein (ECD, alpha IIIb, His Tag), HPLC-verified
Supplier:  Sino Biological 16486H08H1
Encompass_Preferred
A DNA sequence encoding the human FGFR3(alpha(IIIb)) (NP_001156685.1) (Met1-Gly377) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-240-8400