Learn More
Sino Biological Recombinant Human FGFR4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10538H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Asp 369) of human FGFR4 (NP_002002.3) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6792