Learn More
Sino Biological Recombinant Human FGFR4 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10538H08HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Asp369) of human FGFR4 (NP_002002.3) was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5268