Learn More
BIOHIPPO Human FGg Microsample
SDP
Catalog No. 5000314540
Encompass
Human FGg Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FGg in plasma It provides a detection range of 6 25-400 ng/mL with a sensitivity of 2 87 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FGg Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FGg Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FGg biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14540 Supplier BIOHIPPO Supplier No. BHE15211729
Human FGg Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FGg in plasma It provides a detection range of 6 25-400 ng/mL with a sensitivity of 2 87 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FGg Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FGg Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FGg biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color