Learn More
Bon Opus Biosciences Recombinant Human FGL1 (C-hFc) 50ug
SDP
Catalog No. 502172621
Encompass_Preferred
Recombinant Human FGL1 (C-hFc) is produced by Human Cells. The target gene encoding Leu23-Ile312 is expressed with a C-hFc tag. Accession# Q08830. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1019
Catalog No. 50-217-2621 Supplier Bon Opus Biosciences Supplier No. BP101950UG
Recombinant Human FGL1 (C-hFc) is produced by Human Cells. The target gene encoding Leu23-Ile312 is expressed with a C-hFc tag. Accession# Q08830. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1019