Learn More
Bon Opus Biosciences Recombinant Human FGL1 (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP102050UG
Encompass_Preferred
Recombinant Human FGL1 (C-mFc) is produced by Human Cells. The target gene encoding Leu23-Ile312 is expressed with a C-mFc tag. Accession# Q08830. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1020
Catalog No. 50-217-2622