Learn More
Sino Biological Recombinant Human FGL1 Protein (hFc Tag)
Supplier:  Sino Biological 13484H02H150
Encompass_Preferred
A DNA sequence encoding the human FGL1 (NP_004458.3) (Met1-Ile312) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-4233