Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FGL2 Partial ORF (NP_006673, 24 a.a. - 123 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89944548 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-548 25 μg
89-944-547 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-944-548 Supplier Abnova™ Supplier No. H00010875Q01L
Only null left
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a secreted protein that is similar to the beta- and gamma-chains of fibrinogen. The carboxyl-terminus of the encoded protein consists of the fibrinogen-related domains (FRED). The encoded protein forms a tetrameric complex which is stabilized by interchain disulfide bonds. This protein may play a role in physiologic functions at mucosal sites. [provided by RefSeq]

Sequence: NNETEEIKDERAKDVCPVRLESRGKCEEAGECPYQVSLPPLTIQLPKQFSRIEEVFKEVQNLKEIVNSLKKSCQDCKLQADDNGDPGRNGLLLPSTGAPG

Specifications

Accession Number NP_006673
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10875
Molecular Weight (g/mol) 36.74kDa
Name FGL2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen NNETEEIKDERAKDVCPVRLESRGKCEEAGECPYQVSLPPLTIQLPKQFSRIEEVFKEVQNLKEIVNSLKKSCQDCKLQADDNGDPGRNGLLLPSTGAPG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias T49/pT49
Common Name FGL2
Gene Symbol FGL2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.