Learn More
BioVendor, LLC Human Fibrin, Mouse Monoclonal Antibody (100 ug)
SDP
Supplier:  BioVendor, LLC QZBFBY22100
Encompass
Human Fibrin, Mouse Monoclonal Antibody (100 ug). Manufactured by QuickZyme Biosciences. Mouse MAb against human fibrin (also reacts with rabbit, rat, sheep, dog). Reacts with human fibrin, cross-reacts with denatured fibrinogen, does not react with native fibrinogen. SUMMARY: Fibrin functions in hemostasis as a primary component of blood clots. It is also important in the early stages of wound repair. Fibrinolysis is the enzymatic breakdown of fibrin in blood clots and results in various Fibrin Degradation Products (FDP).
Catalog No. 50-206-9107