Learn More
Cayman Chemical Human Fibrinogen (PAD2 Citrullinated), 1 mg
SDP
Supplier:  Cayman Chemical 184731
Encompass_Preferred
Source: Purified from human plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV, citrullinated with human recombinant PAD2 • MW: α chain isoform 1 (95 kDa), α chain isoform 2 (69.8 kDa), β chain (55.9k Da), and isoform γ-B chain (51.5 kDa)
Catalog No. NC1463328