Learn More
Innovative Research Inc Human Fibrinogen ELISA Kit
Supplier:  Innovative Research Inc IHUFBGKT
Encompass_Preferred
This human fibrinogen antigen assay is intended for the quantitative determination of total fibrinogen antigen in human plasma and serum. For research use only. Strip well format. Reagents for up to 96 tests.
Catalog No. 50-203-6111