Learn More
Biomatik Corporation Human Fibroblast Growth Factor 2, Basic (FGF2) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU12137
Encompass_Preferred
Human Fibroblast Growth Factor 2, Basic (FGF2) High-sensitive ELISA Kit for detecting FGF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.47-200pg/ml. Sensitivity: 0.87pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09038. Gene ID: 2247. Target Synonyms: B-FGF; BFGF; FGFB; HBGH-2; Basic Fibroblast Growth Factor; Heparin-binding growth factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5931