Learn More
Biomatik Corporation Human Fibroblast Growth Factor 18 (FGF18) CLIA Kit
Supplier:  Biomatik Corporation EKU12010
Encompass_Preferred
Human Fibroblast Growth Factor 18 (FGF18) CLIA Kit for detecting FGF18 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-500pg/ml. Sensitivity: 13.3pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: O76093. Gene ID: 8817. Target Synonyms: ZFGF5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5299