Learn More
Biomatik Corporation Human Fibroblast Growth Factor 13 (FGF13) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10280
Encompass_Preferred
Human Fibroblast Growth Factor 13 (FGF13) High-sensitive ELISA Kit for detecting FGF13 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.23-100pg/ml. Sensitivity: 0.53pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92913. Gene ID: 2258. Target Synonyms: FHF2; Fibroblast Growth Factor Homologous Factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5339