Learn More
Biomatik Corporation Human Fibroblast Growth Factor Receptor 4 (FGFR4) ELISA Kit
Supplier:  Biomatik Corporation EKU04194
Encompass_Preferred
Human Fibroblast Growth Factor Receptor 4 (FGFR4) ELISA Kit for detecting FGFR4 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P22455. Gene ID: 2264. Target Synonyms: CD334; JTK2; TKF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5942