Learn More
Biomatik Corporation Human Fibroblast Growth Factor 21 (FGF21) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12323
Encompass_Preferred
Human Fibroblast Growth Factor 21 (FGF21) Mini sample ELISA Kit for detecting FGF21 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 2.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NSA1. Gene ID: 26291. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7001