Learn More
Biomatik Corporation Human Fibroblast Growth Factor 11 (FGF11) CLIA Kit
Supplier:  Biomatik Corporation EKU09797
Encompass_Preferred
Human Fibroblast Growth Factor 11 (FGF11) CLIA Kit for detecting FGF11 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.54pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q92914. Gene ID: 2256. Target Synonyms: FHF3; Fibroblast Growth Factor Homologous Factor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7160