Learn More
Biomatik Corporation Human Fibroblast Growth Factor 6 (FGF6) ELISA Kit
Supplier:  Biomatik Corporation EKU04174
Encompass_Preferred
Human Fibroblast Growth Factor 6 (FGF6) ELISA Kit for detecting FGF6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 3.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10767. Gene ID: 2251. Target Synonyms: HBGF6; HST2; HST2 Oncogene; Heparin Secretory-Transforming Protein 2; Heparin-Binding Growth Factor 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8978