Learn More
Biomatik Corporation Human Fibroblast Growth Factor 13 (FGF13) CLIA Kit
Supplier:  Biomatik Corporation EKU10823
Encompass_Preferred
Human Fibroblast Growth Factor 13 (FGF13) CLIA Kit for detecting FGF13 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.69-500pg/ml. Sensitivity: 0.28pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q92913. Gene ID: 2258. Target Synonyms: FHF2; Fibroblast Growth Factor Homologous Factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8281