Learn More
American Research Products Inc Human fibroblast growth factor
Supplier:  American Research Products Inc CSBE04550H
Encompass
Human fibroblast growth factor 9 (glia-actiVating factor) (FGF9) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26316