Learn More
American Research Products Inc Human Fibroblast growth factor
Supplier: American Research Products Inc CSBEL008646HU
Human Fibroblast growth factor receptor 3 (FGFR3) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich