Learn More
American Research Products Inc Human Fibroblast growth factor
Catalog No. 5000327296
Encompass
Human Fibroblast growth factor 5 (FGF5) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27296 Supplier American Research Products Inc Supplier No. CSBEL008632HU
Human Fibroblast growth factor 5 (FGF5) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich