Learn More
Biomatik Corporation Human Fibroblast Growth Factor 12 (FGF12) CLIA Kit
Supplier:  Biomatik Corporation EKU09800
Encompass_Preferred
Human Fibroblast Growth Factor 12 (FGF12) CLIA Kit for detecting FGF12 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.53pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P61328. Target Synonyms: FGF12B; FHF1; Fibroblast Growth Factor Homologous Factor 1; Myocyte-Activating Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4534