Learn More
Biomatik Corporation Human Fibroblast Growth Factor 23 (FGF23) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11924
Encompass_Preferred
Human Fibroblast Growth Factor 23 (FGF23) Mini sample ELISA Kit for detecting FGF23 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9GZV9. Gene ID: 8074. Target Synonyms: ADHR; HYPF; HPDR2; PHPTC; Phosphatonin; Tumor-derived hypophosphatemia-inducing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5251