Learn More
ACROBiosystems Human Fibronectin 500ug
Supplier:  ACROBiosystems FINH5113500UG
Encompass_Preferred
Fibronectin fragment premium grade (FIN-H5113) is expressed from E. coli cells. It contains AA Pro 1361 - Ser 1637 & Ala 1812 - Thr 2107 (Accession # P02751-15).
Catalog No. 50-253-5065