Learn More
Sino Biological Human Fibronectin / Fibronectin Fragment 2 Protein (His Tag) 1mg
Supplier:  Sino Biological 10314H08H1
Encompass_Preferred
A DNA sequence encoding the Fragment 2 (Ser607-Pro1265) of human Fibronectin (NP_997647.1) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-6760