Learn More
Advanced Biomatrix Fibronectin, Human, 1.0 mg
Catalog No. NC0925219
Encompass_Preferred
This Fibronectin solution has been purified from human plasma where it is found as a dimer and has a size of 440-500 kDa with two similar subunits (220-250 kDa) linked by two disulfide bonds. Fibronectin’s primary function is related to cell adhesion and attachment to extracellular matrix. Fibronectin is a widely used broad range natural cell attachment factor. Fibronectin’s primary function is related to cell adhesion and attachment to extracellular matrix. Certain domains of Fibronectin have been found to play important roles that involve interactions with collagen, heparin and other cell surface glycosaminoglycans (GAGs). Fibronectin is ideal for coating of surfaces and is typically used at a concentration of 1-5 ug/cm2. Fibronectin is sterile filtered and is supplied as a ready to use solution after thawing and concentration adjustment.
Catalog No. NC0925219 Supplier Advanced Biomatrix Supplier No. 50501MG
This Fibronectin solution has been purified from human plasma where it is found as a dimer and has a size of 440-500 kDa with two similar subunits (220-250 kDa) linked by two disulfide bonds. Fibronectin’s primary function is related to cell adhesion and attachment to extracellular matrix. Fibronectin is a widely used broad range natural cell attachment factor. Fibronectin’s primary function is related to cell adhesion and attachment to extracellular matrix. Certain domains of Fibronectin have been found to play important roles that involve interactions with collagen, heparin and other cell surface glycosaminoglycans (GAGs). Fibronectin is ideal for coating of surfaces and is typically used at a concentration of 1-5 ug/cm2. Fibronectin is sterile filtered and is supplied as a ready to use solution after thawing and concentration adjustment.