Learn More
Biomatik Corporation Human Fibulin 1 (FBLN1) CLIA Kit
Supplier:  Biomatik Corporation EKU09971
Encompass_Preferred
Human Fibulin 1 (FBLN1) CLIA Kit for detecting FBLN1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 41.2-30,000pg/ml. Sensitivity: 14.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P23142. Gene ID: 2192. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5916