Learn More
Invitrogen™ Human Ficolin-2 ELISA Kit
Description
Human Ficolin-2 quantitates human Ficolin-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Ficolin-2.
Specifications
Specifications
| Accession Number | Q15485 |
| Assay Range | 0.123-30 ng/mL |
| Assay Sensitivity | 0.12 ng/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | 37 kDa elastin-binding protein; Collagen/fibrinogen domain-containing protein 2; EBP-37; FCN2; Fcnb; FCNL; ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin); ficolin 2; ficolin B; ficolin-2; Ficolin-B; ficolin-beta; Hucolin; L-ficolin; P35; RP11-263F14.2; Serum lectin p3537 kDa elastin-binding protein; Collagen/fibrinogen domain-containing protein 2; EBP-37; FCN2; Fcnb; FCNL; ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin); ficolin 2; ficolin B; ficolin-2; Ficolin-B; ficolin-beta; Hucolin; L-ficolin; P35; RP11-263F14.2; Serum lectin p35; |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.