Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Ficolin-2 ELISA Kit

Catalog No. EH192RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH192RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH192RBX10 Supplier Invitrogen™ Supplier No. EH192RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human Ficolin-2 quantitates human Ficolin-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Ficolin-2.

L-Ficolin (Ficolin-2) is an innate immunity pattern recognition molecule found in human serum. L-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP), such as carbohydrates (GlcNAc), lipoteichoic acid and acetylated groups thereby facilitating phagocytosis and activation of complement through the lectin route using the same serine proteases as mannose-binding lectin (MBL) named MASP ((MBL)-associated serine protease).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15485
Assay Range 0.123-30 ng/mL
Assay Sensitivity 0.12 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 37 kDa elastin-binding protein; Collagen/fibrinogen domain-containing protein 2; EBP-37; FCN2; Fcnb; FCNL; ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin); ficolin 2; ficolin B; ficolin-2; Ficolin-B; ficolin-beta; Hucolin; L-ficolin; P35; RP11-263F14.2; Serum lectin p3537 kDa elastin-binding protein; Collagen/fibrinogen domain-containing protein 2; EBP-37; FCN2; Fcnb; FCNL; ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin); ficolin 2; ficolin B; ficolin-2; Ficolin-B; ficolin-beta; Hucolin; L-ficolin; P35; RP11-263F14.2; Serum lectin p35;
Gene ID (Entrez) 2220
Gene Symbol FCN2
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target L-Ficolin
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.