Learn More
BIOHIPPO Human FIL1d Microsample
SDP
Catalog No. 5000314218
Encompass
Human FIL1d Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FIL1d in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 3-2000 pg/mL with a sensitivity of 18 5 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FIL1d Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FIL1d Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FIL1d biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14218 Supplier BIOHIPPO Supplier No. BHE15211968
Human FIL1d Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FIL1d in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 3-2000 pg/mL with a sensitivity of 18 5 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FIL1d Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FIL1d Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FIL1d biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color